Caps for Basik™ 13 mm Screw Top Autosampler Vials, MicroSolv
# de catalogue 97055-360
Fournisseur: MicroSolv Technology Corp
Spécifications
- Cap Size:13-425
- Cap Type:Screw cap
- Septum:Fitted Red Silicone/PTFE
- Material:PP
- Cap Color:Black
- Supplier No.:9504S-05C-BLK