ContactUsLinkComponent
Decapper Tool
Decapper for Vials, MicroSlov
# de catalogue 97054-996
Decapper Tool
Decapper for Vials, MicroSlov
# de catalogue 97054-996
Spécifications
  • Description:
    Decapper Tool
  • Cat. no.:
    97054-996
  • Seal Size:
    20 mm
  • Supplier No.:
    95025-DC
Spécifications
A propos de cet article
These decapper tools are used for detaching aluminum Headspace Caps.
Customers Who Bought This Also Bought