Simplus* Brand Bare Fused Silica Capillary
# de catalogue 97054-824
Fournisseur: MicroSolv Technology Corp
Simplus* Brand Bare Fused Silica Capillary
# de catalogue 97054-824
Fournisseur: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$194.14 to $694.12
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
$377.44 to $992.90
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM®, super gradient grade for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
$741.97 to $47,885.70
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Fournisseur: VWR International
$109.33 to $364.87