Basik*10-425mm Screw-Thread Autosampler Vials
# de catalogue 97054-576
Fournisseur: MicroSolv Technology Corp
Basik*10-425mm Screw-Thread Autosampler Vials
# de catalogue 97054-576
Fournisseur: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$170.25 to $676.42
Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
$599.10 to $2,396.15
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Fournisseur: VWR International
$131.08 to $405.43