ContactUsLinkComponent
Caps
Caps for Autosampler Vials, Screw Top, MicroSolv™/Basik™
# de catalogue 97052-806
Caps
Caps for Autosampler Vials, Screw Top, MicroSolv™/Basik™
# de catalogue 97052-806
Spécifications
  • Cap Size:
    9-425 mm
  • Cap Type:
    Silicone/PTFE Septa
  • Description:
    Green Polypropylene with white silicone
  • Cat. no.:
    97052-806
  • Cap Color:
    Green
  • Supplier No.:
    9502S-E-35G-M