AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
Catalog # 97052-478
Supplier: MicroSolv Technology Corp
Specifications
- Cap size:9-425 mm
- Cap Type:Screw cap, solid, single injection use with a thinned penetration point
- Septum:—
- Material:PE
- Cat. no.:97052-478
- Cap Color:Clear
- Supplier No.:9502S-09-PE