Screw Caps, MicroSolv™
# de catalogue 97051-422
Fournisseur: MicroSolv Technology Corp
Spécifications
- Cap Size:9-425 mm
- Cap Type:Screw cap, knurled
- Septum:Bonded Silicone/PTFE, low durometer
- Material:PP
- Cat. no.:97051-422
- Cap Color:Blue
- Supplier No.:9502S-10C-B
Spécifications
A propos de cet article
These blue screw caps are designed for use with 9-425, 12x32 mm vials.
Customers Who Bought This Also Bought