ContactUsLinkComponent
Screw Cap
Screw Caps with Partial Slit Septa for 12×32 mm Vials, Microsolv Technology Corp
# de catalogue 97051-404
Screw Cap
Screw Caps with Partial Slit Septa for 12×32 mm Vials, Microsolv Technology Corp
# de catalogue 97051-404
Spécifications
  • Color:
    Blue
  • Description:
    Polypropylene Screw Cap with Silicone/PTFE Septa for 12 x 32 mm Vials, 9-425 Thread
  • Cat. no.:
    97051-404
  • Supplier No.:
    9502S-34C-B
Spécifications
A propos de cet article
Screw Caps have a partial precut Y shape slit to prevent evaporation of volatile samples before injection
Helps to minimize vacuum forming in the vial.
Customers Who Bought This Also Bought