ContactUsLinkComponent
Snap Cap, 11Mm Black Color Cap With Starburst PTFE/Silicone Septa
WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
# de catalogue 97050-415
Snap Cap, 11Mm Black Color Cap With Starburst PTFE/Silicone Septa
WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
# de catalogue 97050-415
Spécifications
  • Septum:
    PTFE/silicone, starburst, pre-cut
  • Description:
    Snap cap
  • Cat. no.:
    97050-415
  • Cap Color:
    Black
  • Supplier No.:
    11-0056K
Spécifications
A propos de cet article
These snap-top caps eliminate the need for crimping and decapping tools.
  • Manufactured from PE
  • Fits the WHEATON E-Z Vials® with snap ring top
Available in multiple colors and septa types.
Customers Who Bought This Also Bought