WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
# de catalogue 97050-397
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Septum:PTFE/silicone
- Description:Snap cap, bulk pail
- Cat. no.:97050-397
- Cap Color:Black
- Supplier No.:11-0053K-B
Spécifications
A propos de cet article
These snap-top caps eliminate the need for crimping and decapping tools.
- Manufactured from PE
- Fits the WHEATON E-Z Vials® with snap ring top
Available in multiple colors and septa types.
Customers Who Bought This Also Bought