ContactUsLinkComponent
Seal, 20mm AlumiTin with Aluminum/Silicone Septa
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
# de catalogue 97047-968
Seal, 20mm AlumiTin with Aluminum/Silicone Septa
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
# de catalogue 97047-968
Spécifications
  • Septum:
    Aluminum/Silicone
  • Description:
    AlumiTin™ Large Opening
  • Cat. no.:
    97047-968
  • Cap Color:
    Green
  • Supplier No.:
    20-0060AT
Spécifications
A propos de cet article
MicroLiter's 20 mm caps are available in multiple colors and septa types.
MicroLiter offers products for both LC and GC applications.
Customers Who Bought This Also Bought