WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
# de catalogue 97047-964
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Septum:Aluminum/silicone
- Description:Septa
- Cat. no.:97047-964
- Supplier No.:20-0060
Spécifications
A propos de cet article
These septa are used for 20 mm headspace vial crimp seals.
Designed for optimum performance in a variety of applications and heat ranges.
Customers Who Bought This Also Bought