ContactUsLinkComponent
Seal, 20mm Tear Away with Tan PTFE/White Silicone Septa 0.130
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
# de catalogue 97047-962
Seal, 20mm Tear Away with Tan PTFE/White Silicone Septa 0.130
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
# de catalogue 97047-962
Spécifications
  • Septum:
    Tan PTFE/White Silicone
  • Description:
    Tear-Away Crimp Seals
  • Cat. no.:
    97047-962
  • Cap Color:
    Silver
  • Supplier No.:
    20-0051T
Spécifications
A propos de cet article
MicroLiter's 20 mm caps are available in multiple colors and septa types.
MicroLiter offers products for both LC and GC applications.
Customers Who Bought This Also Bought