ContactUsLinkComponent
Seal, 20 mm Pressure Release with Tan PTFE/White Silicone Septa 0.130
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Catalog # 97047-960
Seal, 20 mm Pressure Release with Tan PTFE/White Silicone Septa 0.130
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Catalog # 97047-960
Specifications
  • Septum:
    Tan PTFE/White Silicone
  • Description:
    Pressure Release Crimp Seals
  • Cat. no.:
    97047-960
  • Cap Color:
    Silver
  • Supplier No.:
    20-0051P
Specifications
About this item
MicroLiter's 20 mm caps are available in multiple colors and septa types.
MicroLiter offers products for both LC and GC applications.
Customers Who Bought This Also Bought