WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
# de catalogue 97047-956
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Septum:Tan PTFE/White Silicone
- Description:Magnetic Crimp Seals, Steel, Large Opening, Standard Apron
- Cat. no.:97047-956
- Cap Color:Gold
- Supplier No.:20-0051MS
Spécifications
A propos de cet article
MicroLiter's 20 mm caps are available in multiple colors and septa types.
MicroLiter offers products for both LC and GC applications.
Customers Who Bought This Also Bought