ContactUsLinkComponent
Septa, 20mm Tan PTFE/White Silicone, 0.130
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Catalog # 97047-940
Septa, 20mm Tan PTFE/White Silicone, 0.130
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Catalog # 97047-940
Specifications
  • Septum:
    Tan PTFE/white silicone
  • Description:
    Septa
  • Cat. no.:
    97047-940
  • Supplier No.:
    20-0051
Specifications
About this item
These septa are used for 20 mm headspace vial crimp seals.
Designed for optimum performance in a variety of applications and heat ranges.
Customers Who Bought This Also Bought