ContactUsLinkComponent
Seal, 20mm AlumiTin with Natural PTFE/Blue Silicone Septa for HS500
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
# de catalogue 97047-918
Seal, 20mm AlumiTin with Natural PTFE/Blue Silicone Septa for HS500
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
# de catalogue 97047-918
Spécifications
  • Septum:
    Natural PTFE/Blue Silicone
  • Description:
    AlumiTin™ Small Opening
  • Cat. no.:
    97047-918
  • Cap Color:
    Green
  • Supplier No.:
    20-0050ATS
Spécifications
A propos de cet article
MicroLiter's 20 mm caps are available in multiple colors and septa types.
MicroLiter offers products for both LC and GC applications.
Customers Who Bought This Also Bought