ContactUsLinkComponent
Septa, 20mm PTFE/Molded Butyl
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Catalog # 97047-892
Septa, 20mm PTFE/Molded Butyl
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Catalog # 97047-892
Specifications
  • Septum:
    Molded PTFE/butyl
  • Description:
    Septa
  • Cat. no.:
    97047-892
  • Supplier No.:
    20-0040
Specifications
About this item
These septa are used for 20 mm headspace vial crimp seals.
Designed for optimum performance in a variety of applications and heat ranges.
Customers Who Bought This Also Bought