ContactUsLinkComponent
Seal, 20mm Magnetic Large Opening Extended Apron with PTFE/Butyl Septa
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Catalog # 97047-880
Seal, 20mm Magnetic Large Opening Extended Apron with PTFE/Butyl Septa
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Catalog # 97047-880
Specifications
  • Septum:
    PTFE/Butyl
  • Description:
    Magnetic Crimp Seals, Steel, Large Opening, Extended Apron
  • Cat. no.:
    97047-880
  • Cap Color:
    Silver
  • Supplier No.:
    20-0030ML
Specifications
About this item
MicroLiter's 20 mm caps are available in multiple colors and septa types.
MicroLiter offers products for both LC and GC applications.
Customers Who Bought This Also Bought