ContactUsLinkComponent
Seal, 20mm Aluminum Standard Apron with PTFE/Butyl Septa
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
# de catalogue 97047-870
Seal, 20mm Aluminum Standard Apron with PTFE/Butyl Septa
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
# de catalogue 97047-870
Spécifications
  • Septum:
    PTFE/Butyl
  • Description:
    Aluminum Crimp Seal
  • Cat. no.:
    97047-870
  • Cap Color:
    Silver
  • Supplier No.:
    20-0030AS
Spécifications
A propos de cet article
MicroLiter's 20 mm caps are available in multiple colors and septa types.
MicroLiter offers products for both LC and GC applications.
Customers Who Bought This Also Bought