ContactUsLinkComponent
Septa, 20mm PTFE/Butyl
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
# de catalogue 97047-866
Septa, 20mm PTFE/Butyl
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
# de catalogue 97047-866
Spécifications
  • Septum:
    PTFE/butyl
  • Description:
    Septa
  • Cat. no.:
    97047-866
  • Supplier No.:
    20-0030
Spécifications
A propos de cet article
These septa are used for 20 mm headspace vial crimp seals.
Designed for optimum performance in a variety of applications and heat ranges.
Customers Who Bought This Also Bought