ContactUsLinkComponent
Seal, 20mm Aluminum Standard Apron, Blue
WHEATON® MicroLiter 20 mm Crimp Seals, DWK Life Sciences
# de catalogue 97047-818
Seal, 20mm Aluminum Standard Apron, Blue
WHEATON® MicroLiter 20 mm Crimp Seals, DWK Life Sciences
# de catalogue 97047-818
Spécifications
  • Description:
    Crimp Seals
  • Cat. no.:
    97047-818
  • For Use With:
    All but LEAP / CTC
  • Cap Color:
    Blue
  • Supplier No.:
    20-0000B