ContactUsLinkComponent
Seal, 20mm Aluminum Standard Apron
WHEATON® MicroLiter 20 mm Crimp Seals, DWK Life Sciences
# de catalogue 97047-816
Seal, 20mm Aluminum Standard Apron
WHEATON® MicroLiter 20 mm Crimp Seals, DWK Life Sciences
# de catalogue 97047-816
Spécifications
  • Description:
    Crimp Seals
  • Cat. no.:
    97047-816
  • For Use With:
    All but LEAP / CTC
  • Cap Color:
    Silver
  • Supplier No.:
    20-0000AS