WHEATON® MicroLiter Screw Caps for Vials, DWK Life Sciences
Catalog # 97047-716
Supplier: MicroLiter Analytical Supply
Specifications
- Cap size:15-425
- Cap Type:Solid top
- Septum:PTFE
- Description:Caps
- Material:Phenolic
- Cat. no.:97047-716
- Cap Color:Black
- Supplier No.:15-0010KPS
Specifications
About this item
These screw-thread closures are available in different configurations.
Customers Who Bought This Also Bought