ContactUsLinkComponent
Cap, Black Solid Top, 15-425 Phenolic With Ptfe Liner
WHEATON® MicroLiter Screw Caps for Vials, DWK Life Sciences
Catalog # 97047-716
Cap, Black Solid Top, 15-425 Phenolic With Ptfe Liner
WHEATON® MicroLiter Screw Caps for Vials, DWK Life Sciences
Catalog # 97047-716
Specifications
  • Cap size:
    15-425
  • Cap Type:
    Solid top
  • Septum:
    PTFE
  • Description:
    Caps
  • Material:
    Phenolic
  • Cat. no.:
    97047-716
  • Cap Color:
    Black
  • Supplier No.:
    15-0010KPS
Specifications
About this item
These screw-thread closures are available in different configurations.
Customers Who Bought This Also Bought