WHEATON® MicroLiter Sample Vial Assembled Kits, DWK Life Sciences
# de catalogue 97047-658
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap included:Yes
- Cap Type:Open top
- Capacity:4.00 ml
- Septum:PTFE
- Color:Clear
- Description:Vial with patch
- Cat. no.:97047-658
- Cap Color:Black
- Supplier No.:13-1210K-A
Spécifications
A propos de cet article
These assembled kits are shipped with the septa installed in the cap and the cap screwed on the vial.
Available in clear and amber formats, with polypropylene or phenolic closures and a variety of septa and liner styles.
Customers Who Bought This Also Bought