ContactUsLinkComponent
Snap Cap, 11Mm Red Color Cap With Precut PTFE/Silicone/ Septa
WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
# de catalogue 97047-206
Snap Cap, 11Mm Red Color Cap With Precut PTFE/Silicone/ Septa
WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
# de catalogue 97047-206
Spécifications
  • Septum:
    PTFE/silicone, pre-cut
  • Description:
    Snap cap
  • Cat. no.:
    97047-206
  • Cap Color:
    Red
  • Supplier No.:
    11-0054R
Spécifications
A propos de cet article
These snap-top caps eliminate the need for crimping and decapping tools.
  • Manufactured from PE
  • Fits the WHEATON E-Z Vials® with snap ring top
Available in multiple colors and septa types.
Customers Who Bought This Also Bought