WHEATON® MicroLiter 11 mm Crimp-Top Seals, DWK Life Sciences
# de catalogue 97047-072
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap Size:11 mm
- Septum:FEP/Butyl Rubber
- Description:Yellow AlumiTin Crimp Seals
- Cat. no.:97047-072
- Supplier No.:11-0021YT
Spécifications
A propos de cet article
MicroLiter's 11 mm crimp-top seals are available in multiple colors and septa types.
Customers Who Bought This Also Bought