Limited Volume Vial Inserts, MicroLiter
# de catalogue 97047-026
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap included:No
- Capacity:0.40 ml
- Description:Silanized insert
- Material:Borosilicate glass
- Bottom Style:Flat bottom
- Supplier No.:11-0000-300S
Spécifications
A propos de cet article
Inserts are made from Type 1 borosilicate glass or polypropylene and meet strict tolerances.
- Conical and flat bottom
- Meet strict tolerances
Conical inserts, inserts with plastic springs, and flat bottom inserts are available to assist in consistent sampling of low-volume samples.
Customers Who Bought This Also Bought