WHEATON® MicroLiter Screw-Thread Vials, 10-425, Component Kits, DWK Life Sciences
# de catalogue 97046-988
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap included:Yes
- Capacity:2.00 ml
- Septum:PTFE/silicone
- Color:Clear
- Description:Vials with patch
- Thickness:60 mil
- Cat. no.:97046-988
- Cap Color:White
- Supplier No.:10-1260W-A
Spécifications
A propos de cet article
The original wide-opening vial available for chromatography.
- Wide-opening
- Multiple configurations available
For applications where the vial remains stationary, this vial is safe to use, even with inserts. MicroLiter stocks all of the popular septa configurations and convenience kits.
Customers Who Bought This Also Bought