ContactUsLinkComponent
9mm Black ScrewTin Cap with PTFE/Silicone/PTFE Septa
Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
# de catalogue 97046-296
9mm Black ScrewTin Cap with PTFE/Silicone/PTFE Septa
Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
# de catalogue 97046-296
Spécifications
  • Ø:
    9 mm
  • Septum:
    ScrewTin™ PTFE/Silicone/PTFE
  • Description:
    Black Polypropylene
  • Cat. no.:
    97046-296
  • Supplier No.:
    09-0040KT
Spécifications
A propos de cet article
MicroLiter's 9 mm screw-thread caps are available in multiple colors and septa types.
Customers Who Bought This Also Bought