Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
# de catalogue 97046-278
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Ø:9 mm
- Septum:Bonded PTFE/Silicone
- Description:Blue Polypropylene
- Cat. no.:97046-278
- Supplier No.:09-0036B
Spécifications
A propos de cet article
MicroLiter's 9 mm screw-thread caps are available in multiple colors and septa types.
Customers Who Bought This Also Bought