ContactUsLinkComponent
9mm Blue Screw Thread Cap Tefzel/Burnt Red Silicone Septa
Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
# de catalogue 97046-220
9mm Blue Screw Thread Cap Tefzel/Burnt Red Silicone Septa
Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
# de catalogue 97046-220
Spécifications
  • Ø:
    9 mm
  • Septum:
    Tefzel®/Silicone
  • Description:
    Blue Polypropylene
  • Supplier No.:
    09-0022B
Spécifications
A propos de cet article
MicroLiter's 9 mm screw-thread caps are available in multiple colors and septa types.
Customers Who Bought This Also Bought