ContactUsLinkComponent
Septa PTFE 0.010 for 9mm Cap
Septa for 9 mm Screw-Thread Caps, MicroLiter
# de catalogue 97046-176
Septa PTFE 0.010 for 9mm Cap
Septa for 9 mm Screw-Thread Caps, MicroLiter
# de catalogue 97046-176
Spécifications
  • Ø:
    9 mm
  • Septum:
    PTFE
  • Description:
    Septa
  • Cat. no.:
    97046-176
  • Supplier No.:
    09-0010
Spécifications
A propos de cet article
MicroLiter's 9 mm screw-thread caps are available in multiple colors and septa types.
  • ScewTin™ Ring is a metallic disc for x-y-z robotics that employ magnetic capture to move vials
  • Post Baked to remove additional siloxanes
Customers Who Bought This Also Bought