Caps and Seals for 8 mm Crimp Top Vials, Snap Caps, and Aluminum Seals, MicroLiter, DWK Life Sciences
# de catalogue 97046-020
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Septum:PTFE/Silicone/PTFE
- Description:Aluminum Crimp Seal
- Cat. no.:97046-020
- Supplier No.:08-0040A