ContactUsLinkComponent
Kit, Natural Snap Cap With PTFE/Silicone Stopper And 2.2Ml Round Inserts
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
# de catalogue 97044-802
Kit, Natural Snap Cap With PTFE/Silicone Stopper And 2.2Ml Round Inserts
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
# de catalogue 97044-802
Spécifications
  • Septum:
    PTFE/Silicone Stopper
  • Well Volume:
    2.2 mL
  • Color:
    Clear
  • Material:
    PP
  • Cat. no.:
    97044-802
  • Cap Color:
    Natural
  • Well Bottom Shape:
    Round
  • Supplier No.:
    07-0045N2200
Spécifications
A propos de cet article
MicroLiter µLPlate® components include a proprietary product line of 96-well round and square well microplates with glass inserts and color-coded caps.
  • Variety of volumes available
  • Standard chromatography closures
  • Caps with septa
  • Silicone mats are resealable
A comprehensive line of 96-Well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials
Polypropylene μLPlates® have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. Users may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system allows even GC samples to be converted to 96-Well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/Silicone rubber using a mechanical seal. A variety of insert options allows users to choose the specifications appropriate for any application.
The system combines microplates, glass inserts, μLmats™, and caps with chromatography-based septa to offer secure vessels to maintain even the most volatile samples.
Customers Who Bought This Also Bought