ContactUsLinkComponent
Vwr* Foam Insert
Accessories for VWR® Standard and Advanced Heavy-Duty Vortex Mixers
Catalog # 97043-590
Supplier:  VWR International
Product available on GSA Advantage®
   Waste Reducing
undefined
Accessories for VWR® Standard and Advanced Heavy-Duty Vortex Mixers
Catalog # 97043-590
Supplier:  VWR International
Product available on GSA Advantage®
   Waste Reducing
Specifications
  • Description:
    VWR® Foam Tube Insert for Twenty 15–18 mm Dia. Test Tubes (Requires Universal Vessel Holder)
  • Environmentally Preferable:
    Waste Reducing
  • Cat. no.:
    97043-590