HIGH TEMP FOOD PASTEURIZATION
# de catalogue 97017-222
Fournisseur: Cryopak Verification Technologies
HIGH TEMP FOOD PASTEURIZATION
# de catalogue 97017-222
Fournisseur: Cryopak Verification Technologies
Customers Who Bought This Also Bought
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Fournisseur: MilliporeSigma
2 699,72 $
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
64,40 $ to 734,93 $
Potassium dihydrogen phosphate, anhydrous ≥98% NF, Ph. Eur. endotoxin tested
Fournisseur: Avantor Performance Materials
489,50 $
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
377,44 $ to 992,90 $