To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Ø:12 mm
- Accuracy:0.03 pH for Buffers using ATC
- Length:120 mm
- Description:pH/ATC Triode™ combination electrodes, with BNC waterproof, 8-Pin Mini DIN connector
- Operating temperature range:0 to 80°C
- Body material:Epoxy body filled with low maintenance gel
- Cat. no.:97015-740
- Cable Length:3 m
- Supplier No.:8107UWMMD
Spécifications
A propos de cet article
Increase the accuracy of readings with a pH sensor that delivers superior performance at varying sample temperatures. Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ combination electrodes combine the stability, precision, and speed of response of the ROSS Ultra™ line with integrated temperature measurement.
- Measure pH and Temperature with one low-maintenance electrode
- Compatible with Orion™ Versa Star™, Orion Star™ A, and Orion™ Dual Star™ meters
- Ideal for pharmaceutical/biopharmaceutical, food manufacturing, academic/government, and environmental applications
- Glass capillary junction
- Isopotential point: 7
- pH range of 0 to 14
Low ionic strength samples, such as clean waters, present a challenge to pH electrode performance.
Complete pH measurements faster, with the assurance of a long, maintenance- free electrode lifetime due to the special long-life gel and innovative capillary glass junction design. Maintain sample integrity, even in TRIS buffers, as these double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for routine sample measurements.
Sluggish response, poor accuracy, and drift are common. These electrodes respond within one minute, providing a stable and accurate reading.