To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Large Display ATC pH Pen
- Model:Large Display ATC model
- Dimensions:165 × 38 × 38 mm (6.5 × 1.5 × 1.5")
- Temperature compensation:Automatic
- Weight:2.5 oz (68 g)
- pH accuracy:±0.2
- pH calibration:3 points
- pH range:0 to 14
- pH resolution:0.1
Spécifications
A propos de cet article
Easy to carry, simple to operate and sufficient for most field pH tests.
- Highly stable and accurate readings with a large LCD display
- Designed for simple one-hand operation
- Features hold function, and auto power off with low battery indicator
Automatically calibrates to pH 4, 7, 10 buffers and can be adjusted to recognize non-standard buffers as well. The case is IP65 waterproof, and will float if accidentally dropped into the water.
Large Display pH Pen: Has a Range of 2 to 12 with an Accuracy of ±0.3 pH and is the perfect instrument for economical spot sampling of pH.
Large Display ATC pH Pen: Has a Range of 0 to 14 with an Accuracy of ±0.2 pH and is the right instrument for spot sampling of pH and temperature. Displays temperature in both °C and °F.
Large Display ATC 0.01 pH Pen: Has a Range of 0 to 14 with an Accuracy of ±0.05 pH and is the ideal instrument when you need laboratory accuracy in the field for sampling of pH and temperature. Displays temperature in both °C and °F.