VWR® CryoGuard Cryogenic Gloves
Product Details & Documents
Multi-layer, insulated gloves are lightweight, flexible, and durable
- Multi-layer and insulated
- Lightweight, flexible, and durable
- Moisture-wicking liner
- Waterproof
- Various lengths available
Multi-layer, insulated gloves are designed to protect hands and arms from the hazards of working with cryogenic materials. They can be worn for extended periods of time, as they are extremely warm, even in temperatures as low as –196°C. Water-resistant and waterproof gloves feature two layers of insulation material bonded together at the edges to trap insulating air without adding additional bulk and an inner lining to wick away moisture from the skin. Waterproof gloves incorporate a thin, seamless, 100% waterproof inner protective layer. All wrist-length gloves also incorporate an uninsulated knit cuff.
These gloves allow for dexterity when working in temperatures as low as –196°C. Two layers of insulation material, bonded together at the edges, traps air without adding additional bulk. Inner lining wicks moisture from the skin.
Ordering information: Gloves are not intended for immersion in liquid nitrogen. Gloves are packed one pair per each unit.
Spécifications
- Glove Color:Blue
- Taille du gant:M
- Longueur:Mid-arm