Fluff Underpad, DUKAL™ Corporation
Catalog # 95041-734
Supplier: Dukal
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Nonsterile Fluff Underpad
- Dimensions:17×24"
- Cat. no.:95041-734
- Supplier No.:11724
Specifications
About this item
Highly absorbent with a cellulose fill that does not shift during use.
- Soft spunbonded facing and blue polypropylene backing
- Vacuum packed for maximum storage efficiency
- Not made with natural rubber latex
Packaging: Packaged 100 per bag, 3 bags per case.
Customers Who Bought This Also Bought