AccuTarget™ Control siRNA Set, Bioneer
Catalog # 95030-562
Supplier: Bioneer
CAS Number:
Specifications
- Description:AccuTarget™ Negative Control siRNA
- Cat. no.:95030-562
- Purification Type:BioRP
- Scale:10 nmole
- Supplier No.:SN-1002
Specifications
About this item
AccuTarget™ Control siRNAs are a wide range of positive control siRNAs, negative control siRNAs and control siRNA sets
Control siRNA can be fluorescently labeled for monitoring transfection efficiency.
Customers Who Bought This Also Bought