Spécifications
- Capacity:120 ml (4 oz.)
- Cat. no.:95025-622
- Supplier No.:ES3700
Spécifications
A propos de cet article
Pathology containers are designed for collection, transport, and storage of powders, liquids, solids, and biological specimens.
- Leak resistant seal
- Durable PP construction
- Non sterile
- Snap-top lids
All containers are nonsterile and feature snap-top lids that provide a secure, leak-resistant seal.