Safety Cone Signs, National Marker
# de catalogue 94008-118
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Cat. no.:94008-118
- Legend:No Parking Graphic
- Dimensions:26.4cm×32.1cm (10⅜×12⅝")
- Supplier No.:CS11
Spécifications
A propos de cet article
Install temporary signage quick and easy.
0.125 expanded PVC 103/8 x 125/8"
Specially designed cut-grooves for easy installation
Weather resistant
One sided message
Customers Who Bought This Also Bought