Spécifications
- Description:Heavy weight
- Packaging:100 Pads
- Dimensions:25.4x33 cm (10x13)"
- Supplier No.:MAT251
Spécifications
A propos de cet article
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated.
- Eight-layer pads and rolls combine grip and strength to reduce slip hazards and keep floors clean
- Dispenser box makes it easy to take, carry and store mats while protecting from dirt and moisture
Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
Absorbs and retains oils, coolants, solvents and water. Dark gray color hides grime so mat stays on the job longer. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Can be wrung out and incinerated after use to reduce waste or for fuels blending. Dispenser box makes it easy to take, carry and store your mat while protecting it from dirt and moisture. Pads are ideal for catching drips and soaking up spills. Easy-tear perforations let you take only what you need so you use less mat and save money.
Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups. Mediumweight construction offers great performance for everyday tasks and cleanups.
Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind.