ContactUsLinkComponent
Accessories for VWR® Heated Circulating Baths
Catalogue # 89201-010
Fournisseur:  VWR International
Product available on GSA Advantage®
undefined
Accessories for VWR® Heated Circulating Baths
Catalogue # 89201-010
Fournisseur:  VWR International
Product available on GSA Advantage®
Spécifications
  • Description:
    RS232 Cable
  • Cat. no.:
    89201-010
  • For Use With:
    VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable Controllers
  • Details:
    Cable Length: 9'