Accessories for VWR® Heated Circulating Baths
Catalogue # 89201-010
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Description:RS232 Cable
- Cat. no.:89201-010
- For Use With:VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable Controllers
- Details:Cable Length: 9'
Spécifications
À propos de cet article
Frequently Bought Together