ContactUsLinkComponent
Matrix* 384-Channel Automation Reservoir
Disposable Automation Reservoirs
# de catalogue 89200-086
Disposable Automation Reservoirs
# de catalogue 89200-086
Spécifications
  • Description:
    384 channel
  • Supplier No.:
    1064-05-7
Spécifications
A propos de cet article
Disposable automation reservoirs are of polypropylene construction and are designed for simple, economical handling of reagents on automated systems.
The standard microplate dimensions enables reservoirs to be used in plate handlers or stackers.
With the same dimensions as standard microplates, these disposable reservoirs can be used in plate handlers or stackers. The removable splash baffles prevent reagents from splashing out when being moved on robotic decks. They are compatible with virtually any automated liquid handler as well as all of our automated pipette tips.
Customers Who Bought This Also Bought