To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Cap included:Yes
- Capacity:45.00 ml
- Description:Tamper Evident Vial
- Cat. no.:89170-922
- Supplier No.:74829
Spécifications
A propos de cet article
Leak-tight 45 ml tamper evident vials are closed in the mold during manufacturing to form a hermetic seal, including a plastic filament that connects the hinged lid to the base of the vial.
Once opened, the plastic filament will separate, indicating the first time the vial has been opened since leaving the mold. Each vial is graduated at the 15 and 30 ml capacity level and the virgin polypropylene enables clear visibility into the collected specimen container. The 45 ml is the industry choice for secure collection and transport.