Spécifications
- Cap Size:120 mm
- Capacity:20 L (5.3 gal.)
- Cat. no.:89170-772
Spécifications
A propos de cet article
VWR® HDPE carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use.
- Chemical-resistant
- HDPE construction
- Molded grips for easy lifting, handling, and pouring
The distinctive rectangular shape saves valuable bench space.
VWR® HDPE carboys are molded from a translucent USP Class VI, and FDA grade resin. They offer excellent chemical resistance, making them ideal for most buffers and reagents as well as powders and solids. Versatile caps, adapters, versatile barbs, and spigots are constructed of polypropylene. The recommended autoclaving cycle for versatile caps, adapters, spigots, and versatile barbs is 20 minutes at 15 psi and 121 °C (250 °F).
Made in the USA.
Large handles on top of the container and molded handles in the base provide a full, secure grip to make lifting, carrying, and pouring easy. Specified models are available with a 120 mm wide neck, which allows the user to reach inside the carboy for easier cleaning. Leakproof polypropylene versatile caps offer the ultimate in connection flexibility. Open caps (sold separately) are compatible with interchangeable adapters, which feature a wide variety of top-connections, including molded-in hose barbs and quick-connect hose barbs. All carboys have their material and volume clearly imprinted on the outside of the bottle, as well as large, easy to read metric graduation marks which are certified to ±5% accuracy.