ContactUsLinkComponent
VWR's Fly Lock Plugs
VWR® Drosophila Vial Plugs, Cellulose Acetate
# de catalogue 89168-890
Fournisseur:  VWR International
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
VWR's Fly Lock Plugs
VWR® Drosophila Vial Plugs, Cellulose Acetate
# de catalogue 89168-890
Fournisseur:  VWR International
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Spécifications
  • Description:
    Fits Bottles
Spécifications
A propos de cet article
Bonded dense-weave cellulose acetate plugs provide a better barrier to mites with minimal shedding.
  • Cellulose acetate fabrication
  • Excellent barrier to mites
  • Minimal shedding
Reduce evaporation to keep food fresh longer.
Customers Who Bought This Also Bought