VWR® Drosophila Vial Plugs, Cellulose Acetate
# de catalogue 89168-890
Fournisseur: VWR International
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Spécifications
- Description:Fits Bottles
Spécifications
A propos de cet article
Bonded dense-weave cellulose acetate plugs provide a better barrier to mites with minimal shedding.
- Cellulose acetate fabrication
- Excellent barrier to mites
- Minimal shedding
Reduce evaporation to keep food fresh longer.
Customers Who Bought This Also Bought